missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 15419135
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 325461-35-6
Molecular Formula C28H46O4
Molecular Weight (g/mol) 446.672
MDL Number MFCD03427237
InChI Key JMACLBZCZVVLDQ-UHFFFAOYSA-N
Synonym 2,5-bis 3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctoxy terephthalaldehyde, 2,5-bis-3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctyl oxy benzene-1,4-dicarbaldehyde
PubChem CID 3653721
IUPAC Name 2,5-bis(3,7-dimethyloctoxy)terephthalaldehyde
SMILES CC(C)CCCC(C)CCOC1=CC(=C(C=C1C=O)OCCC(C)CCCC(C)C)C=O

Specifications

Quantity 1 g
Formula Weight 446.66
Percent Purity 98%
Chemical Name or Material 2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'