Learn More
2,4-Dichlorobenzaldehyde, 98%, Thermo Scientific Chemicals
30.20 € - 103.00 €
Chemical Identifiers
CAS | 874-42-0 |
---|---|
Molecular Formula | C7H4Cl2O |
Molecular Weight (g/mol) | 175.01 |
MDL Number | MFCD00003305 |
InChI Key | YSFBEAASFUWWHU-UHFFFAOYSA-N |
Synonym | benzaldehyde, 2,4-dichloro, unii-z08ql2124r, ccris 6013, 2,4-dichloro benzaldehyde, 2,4-dichloro-benzaldehyde, acmc-209qnb, dsstox_cid_2130, 2,4 dichloro benzaldehyde, 2,4-di-chlorobenzaldehyde, dsstox_rid_76500 |
PubChem CID | 13404 |
IUPAC Name | 2,4-dichlorobenzaldehyde |
SMILES | C1=CC(=C(C=C1Cl)Cl)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10205550
|
Thermo Scientific Acros
164140050 |
5 g | Glass bottle |
30.20 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10625032
|
Thermo Scientific Acros
164141000 |
100 g | Plastic bottle |
31.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10489070
|
Thermo Scientific Acros
164145000 |
500 g | Plastic bottle |
103.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
874-42-0 | |
175.01 | |
YSFBEAASFUWWHU-UHFFFAOYSA-N | |
13404 | |
C1=CC(=C(C=C1Cl)Cl)C=O |
C7H4Cl2O | |
MFCD00003305 | |
benzaldehyde, 2,4-dichloro, unii-z08ql2124r, ccris 6013, 2,4-dichloro benzaldehyde, 2,4-dichloro-benzaldehyde, acmc-209qnb, dsstox_cid_2130, 2,4 dichloro benzaldehyde, 2,4-di-chlorobenzaldehyde, dsstox_rid_76500 | |
2,4-dichlorobenzaldehyde |
Specifications
874-42-0 | |
100.0 | |
Yellow to Beige | |
135°C | |
97.5% min. (GC) | |
Cl2C6H3CHO | |
07, 236 | |
Solubility in water: insoluble in water. Other solubilities: soluble in methanol,ethanol,ether and acetone | |
C1=CC(=C(C=C1Cl)Cl)C=O | |
175.01 | |
175.01 | |
Crystalline Powder, Crystals or Chunks |
97.5 | |
69°C to 73°C | |
233°C | |
Authentic | |
C7H4Cl2O | |
MFCD00003305 | |
benzaldehyde, 2,4-dichloro, unii-z08ql2124r, ccris 6013, 2,4-dichloro benzaldehyde, 2,4-dichloro-benzaldehyde, acmc-209qnb, dsstox_cid_2130, 2,4 dichloro benzaldehyde, 2,4-di-chlorobenzaldehyde, dsstox_rid_76500 | |
YSFBEAASFUWWHU-UHFFFAOYSA-N | |
2,4-dichlorobenzaldehyde | |
13404 | |
98% | |
2, 4-Dichlorobenzaldehyde, 98% |
Safety and Handling
GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-861-1
missing translation for 'tsca' : TSCA
- 2,4-Dichlorobenzaldehyde
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only