missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 10489070
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-42-0
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00003305
InChI Key YSFBEAASFUWWHU-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dichloro, unii-z08ql2124r, ccris 6013, 2,4-dichloro benzaldehyde, 2,4-dichloro-benzaldehyde, acmc-209qnb, dsstox_cid_2130, 2,4 dichloro benzaldehyde, 2,4-di-chlorobenzaldehyde, dsstox_rid_76500
PubChem CID 13404
IUPAC Name 2,4-dichlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)Cl)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69°C to 73°C
Color Yellow to Beige
Boiling Point 233°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula Cl2C6H3CHO
Quantity 500 g
Beilstein 07, 236
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in methanol,ethanol,ether and acetone
Formula Weight 175.01
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2, 4-Dichlorobenzaldehyde, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dichlorobenzaldehyde
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'