missing translation for 'onlineSavingsMsg'
Learn More

2',3',4'-Trichloroacetophenone, 95%, Thermo Scientific™

23.70 € - 176.00 €

Chemical Identifiers

CAS 13608-87-2
Molecular Formula C8H5Cl3O
Molecular Weight (g/mol) 223.477
MDL Number MFCD00000542
InChI Key BXJZZJYNVIDEKG-UHFFFAOYSA-N
Synonym 2',3',4'-trichloroacetophenone, 1-2,3,4-trichlorophenyl ethanone, 2,3,4-trichloroacetophenone, ethanone, 1-2,3,4-trichlorophenyl, 1-2,3,4-trichlorophenyl ethan-1-one, ccris 785, unii-8hvd33n785, acetophenone, 2',3',4'-trichloro, ethanone,1-2,3,4-trichlorophenyl, 1-acetyl-2,3,4-trichlorobenzene
PubChem CID 26132
IUPAC Name 1-(2,3,4-trichlorophenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)Cl)Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11424864
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13790.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11434864
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13790.14
25 g
70.20 €
25g
Please to purchase this item. Need a web account? Register with us today!
11444864
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13790.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13608-87-2
223.477
BXJZZJYNVIDEKG-UHFFFAOYSA-N
26132
CC(=O)C1=C(C(=C(C=C1)Cl)Cl)Cl
C8H5Cl3O
MFCD00000542
2',3',4'-trichloroacetophenone, 1-2,3,4-trichlorophenyl ethanone, 2,3,4-trichloroacetophenone, ethanone, 1-2,3,4-trichlorophenyl, 1-2,3,4-trichlorophenyl ethan-1-one, ccris 785, unii-8hvd33n785, acetophenone, 2',3',4'-trichloro, ethanone,1-2,3,4-trichlorophenyl, 1-acetyl-2,3,4-trichlorobenzene
1-(2,3,4-trichlorophenyl)ethanone

Specifications

13608-87-2
C8H5Cl3O
1948674
BXJZZJYNVIDEKG-UHFFFAOYSA-N
1-(2,3,4-trichlorophenyl)ethanone
26132
95%
60°C to 64°C
MFCD00000542
2',3',4'-trichloroacetophenone, 1-2,3,4-trichlorophenyl ethanone, 2,3,4-trichloroacetophenone, ethanone, 1-2,3,4-trichlorophenyl, 1-2,3,4-trichlorophenyl ethan-1-one, ccris 785, unii-8hvd33n785, acetophenone, 2',3',4'-trichloro, ethanone,1-2,3,4-trichlorophenyl, 1-acetyl-2,3,4-trichlorobenzene
CC(=O)C1=C(C(=C(C=C1)Cl)Cl)Cl
223.477
223.49
2',3',4'-Trichloroacetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 237-092-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications