missing translation for 'onlineSavingsMsg'
Learn More

2',3',4'-Trichloroacetophenone, 95%, Thermo Scientific™

Product Code. 11424864
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13608-87-2
Molecular Formula C8H5Cl3O
Molecular Weight (g/mol) 223.477
MDL Number MFCD00000542
InChI Key BXJZZJYNVIDEKG-UHFFFAOYSA-N
Synonym 2',3',4'-trichloroacetophenone, 1-2,3,4-trichlorophenyl ethanone, 2,3,4-trichloroacetophenone, ethanone, 1-2,3,4-trichlorophenyl, 1-2,3,4-trichlorophenyl ethan-1-one, ccris 785, unii-8hvd33n785, acetophenone, 2',3',4'-trichloro, ethanone,1-2,3,4-trichlorophenyl, 1-acetyl-2,3,4-trichlorobenzene
PubChem CID 26132
IUPAC Name 1-(2,3,4-trichlorophenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)Cl)Cl)Cl

Specifications

Melting Point 60°C to 64°C
Quantity 5 g
Beilstein 1948674
Formula Weight 223.49
Percent Purity 95%
Chemical Name or Material 2',3',4'-Trichloroacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'