Learn More
2-(2-Aminoethyl)pyridine, 98%, Thermo Scientific Chemicals
32.40 € - 435.00 €
Chemical Identifiers
CAS | 2706-56-1 |
---|---|
Molecular Formula | C7H10N2 |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00006367 |
InChI Key | XPQIPUZPSLAZDV-UHFFFAOYSA-N |
Synonym | 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine |
PubChem CID | 75919 |
ChEBI | CHEBI:74024 |
SMILES | NCCC1=CC=CC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397515
|
Thermo Scientific Alfa Aesar
L05627.03 |
1 g |
32.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307525
|
Thermo Scientific Alfa Aesar
L05627.06 |
5 g |
105.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317525
|
Thermo Scientific Alfa Aesar
L05627.14 |
25 g |
435.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.
Solubility
Soluble in water (partly), and most common polar solvent. Insoluble in non-polar solvents, and oils.
Notes
Air Sensitive. Store at 4°C. Stor in the dark. Incompatible with air, oxidizing agents, carbondioxide.
Chemical Identifiers
2706-56-1 | |
122.17 | |
XPQIPUZPSLAZDV-UHFFFAOYSA-N | |
75919 | |
NCCC1=CC=CC=N1 |
C7H10N2 | |
MFCD00006367 | |
2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine | |
CHEBI:74024 |
Specifications
2706-56-1 | |
208°C | |
Amine-like | |
1.536 | |
UN2735 | |
Air Sensitive | |
Soluble in water (partly),and most common polar solvent. Insoluble in non-polar solvents,and oils. | |
NCCC1=CC=CC=N1 | |
122.17 | |
CHEBI:74024 | |
98% |
1.02 | |
100°C (212°F) | |
C7H10N2 | |
MFCD00006367 | |
111208 | |
2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine | |
XPQIPUZPSLAZDV-UHFFFAOYSA-N | |
2-pyridin-2-ylethanamine | |
75919 | |
122.17 | |
2-(2-Aminoethyl)pyridine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 220-295-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only