missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 11307525
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.

Solubility
Soluble in water (partly), and most common polar solvent. Insoluble in non-polar solvents, and oils.

Notes
Air Sensitive. Store at 4°C. Stor in the dark. Incompatible with air, oxidizing agents, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2706-56-1
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006367
InChI Key XPQIPUZPSLAZDV-UHFFFAOYSA-N
Synonym 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine
PubChem CID 75919
ChEBI CHEBI:74024
SMILES NCCC1=CC=CC=N1

Specifications

Density 1.02
Boiling Point 208°C
Flash Point 100°C (212°F)
Odor Amine-like
Refractive Index 1.536
Quantity 5 g
UN Number UN2735
Beilstein 111208
Sensitivity Air Sensitive
Solubility Information Soluble in water (partly),and most common polar solvent. Insoluble in non-polar solvents,and oils.
IUPAC Name 2-(pyridin-2-yl)ethan-1-amine
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2-(2-Aminoethyl)pyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'