Learn More
(1S,2S)-Cyclohexane-1,2-dicarboxylic acid, 98+%, Thermo Scientific Chemicals
86.53 € - 334.67 €
Chemical Identifiers
CAS | 21963-41-7 |
---|---|
Molecular Formula | C8H12O4 |
Molecular Weight (g/mol) | 172.18 |
MDL Number | MFCD00151258,MFCD00001464 |
InChI Key | QSAWQNUELGIYBC-UHFFFAOYNA-N |
Synonym | 1s,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, trans, 1,2-cyclohexanedicarboxylic acid, 1s,2s, 1,2-cyclohexanedicarboxylicacid, 1s,2s, pubchem19269, trans-hexahydrophthalic acid, 1alpha,2beta-cyclohexanedicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylicacid |
PubChem CID | 75315 |
IUPAC Name | (1S,2S)-cyclohexane-1,2-dicarboxylic acid |
SMILES | OC(=O)C1CCCCC1C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15439715
|
Thermo Scientific Alfa Aesar
|
250 mg |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478907
|
Thermo Scientific Alfa Aesar
H52290.03 |
1 g |
86.53 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488907
|
Thermo Scientific Alfa Aesar
H52290.06 |
5 g |
334.67 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.
Solubility
Soluble in acetone(almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
21963-41-7 | |
172.18 | |
QSAWQNUELGIYBC-UHFFFAOYNA-N | |
75315 | |
OC(=O)C1CCCCC1C(O)=O |
C8H12O4 | |
MFCD00151258,MFCD00001464 | |
1s,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, trans, 1,2-cyclohexanedicarboxylic acid, 1s,2s, 1,2-cyclohexanedicarboxylicacid, 1s,2s, pubchem19269, trans-hexahydrophthalic acid, 1alpha,2beta-cyclohexanedicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylicacid | |
(1S,2S)-cyclohexane-1,2-dicarboxylic acid |
Specifications
21963-41-7 | |
MFCD00151258,MFCD00001464 | |
Soluble in acetone(almost transparency). | |
OC(=O)C1CCCCC1C(O)=O | |
172.18 | |
172.18 | |
(1S,2S)-Cyclohexane-1,2-dicarboxylic acid |
C8H12O4 | |
1s,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, trans, 1,2-cyclohexanedicarboxylic acid, 1s,2s, 1,2-cyclohexanedicarboxylicacid, 1s,2s, pubchem19269, trans-hexahydrophthalic acid, 1alpha,2beta-cyclohexanedicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylicacid | |
QSAWQNUELGIYBC-UHFFFAOYNA-N | |
(1S,2S)-cyclohexane-1,2-dicarboxylic acid | |
75315 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.