missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-Cyclohexane-1,2-dicarboxylic acid, 98+%, Thermo Scientific Chemicals

Product Code. 15439715
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.

Solubility
Soluble in acetone(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21963-41-7
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00151258,MFCD00001464
InChI Key QSAWQNUELGIYBC-UHFFFAOYNA-N
Synonym 1s,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, trans, 1,2-cyclohexanedicarboxylic acid, 1s,2s, 1,2-cyclohexanedicarboxylicacid, 1s,2s, pubchem19269, trans-hexahydrophthalic acid, 1alpha,2beta-cyclohexanedicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylicacid
PubChem CID 75315
IUPAC Name (1S,2S)-cyclohexane-1,2-dicarboxylic acid
SMILES OC(=O)C1CCCCC1C(O)=O

Specifications

Quantity 250 mg
Solubility Information Soluble in acetone(almost transparency).
Formula Weight 172.18
Percent Purity ≥98%
Chemical Name or Material (1S,2S)-Cyclohexane-1,2-dicarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.