Learn More
10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%, Thermo Scientific Chemicals
101.00 € - 275.00 €
Chemical Identifiers
CAS | 944721-52-2 |
---|---|
Molecular Formula | C20H29F5O5Si |
Molecular Weight (g/mol) | 472.524 |
MDL Number | MFCD13181915 |
InChI Key | KSYPNQIBGIOYPZ-UHFFFAOYSA-N |
Synonym | 11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate |
PubChem CID | 23582157 |
IUPAC Name | (2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate |
SMILES | CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478785
|
Thermo Scientific Alfa Aesar
H34117.MD |
250 mg |
101.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15468785
|
Thermo Scientific Alfa Aesar
H34117.03 |
1 g |
275.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly sealed and place it in a cool, dry and well ventilated place. Store under inert gas.
Chemical Identifiers
944721-52-2 | |
472.524 | |
KSYPNQIBGIOYPZ-UHFFFAOYSA-N | |
23582157 | |
CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC |
C20H29F5O5Si | |
MFCD13181915 | |
11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate | |
(2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate |
Specifications
145°C to 150°C (0.05mmHg) | |
C20H29F5O5Si | |
11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate | |
CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC | |
472.524 | |
472.52 | |
1.4370-1.4410 | |
10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane |
944721-52-2 | |
MFCD13181915 | |
KSYPNQIBGIOYPZ-UHFFFAOYSA-N | |
(2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate | |
23582157 | |
95% | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.