missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly sealed and place it in a cool, dry and well ventilated place. Store under inert gas.
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly sealed and place it in a cool, dry and well ventilated place. Store under inert gas.
Specifications
Specifications
Boiling Point | 145°C to 150°C (0.05mmHg) |
Quantity | 1 g |
Formula Weight | 472.52 |
Percent Purity | 95% |
Refractive Index | 1.4370-1.4410 |
Sensitivity | Moisture sensitive |
Chemical Name or Material | 10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'