missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol, CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haen
CHROMASOLV™, for HPLC,≥99.9%
49.71 € - 1124.88 €
Chemical Identifiers
CAS | 71-23-8 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.10 |
MDL Number | MFCD00002941 |
InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
PubChem CID | 1031 |
ChEBI | CHEBI:28831 |
IUPAC Name | propan-1-ol |
SMILES | CCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15654640
|
Honeywell-Chromasolv
34871-100ML |
100 mL | Glass Bottle | N/A | |||||
15664640
|
Honeywell-Chromasolv
34871-1L |
1 L | Glass Bottle |
151.03 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15684640
|
Honeywell-Chromasolv
34871-2L |
2 L | Glass Bottle | N/A | |||||
15674640
|
Honeywell-Chromasolv
34871-2.5L |
2.5 L | Glass Bottle |
317.24 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15604650
|
Honeywell-Chromasolv
34871-6X1L |
6 x 1 L | Glass Bottle |
808.44 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15694640
|
Honeywell-Chromasolv
34871-4X2.5L |
4 x 2.5 L | Glass Bottle |
1124.88 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
CHEBI:28831 | |
CCCO |
C3H8O | |
MFCD00002941 | |
1031 | |
propan-1-ol |
Specifications
-127°C (literature) | |
100.0000% | |
96°C to 98°C | |
C3H8O | |
MFCD00002941 | |
1098242 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1 | |
HPLC |
71-23-8 | |
0.804 g/mL at 25 °C(lit.) | |
21°C | |
CH3CH2CH2OH | |
UN1274 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CCCO | |
60.10 | |
CHEBI:28831 | |
>99.9% | |
1-Propanol, CHROMASOLV™, for HPLC, ≥99.9% |
Safety and Handling
missing translation for 'shelfLife' : 1620 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: n-Propanol
missing translation for 'einecsNumber' : 200-746-9
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Propanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.