missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haen

Product Code. 15654640
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°C (literature)
CAS Max % 100.0000%
Density 0.804 g/mL at 25 °C(lit.)
Boiling Point 96°C to 98°C
Quantity 100 mL
Flash Point 21°C
Packaging Glass Bottle
Linear Formula CH3CH2CH2OH
UN Number UN1274
Beilstein 1098242
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
Formula Weight 60.1
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material 1-Propanol, CHROMASOLV™, for HPLC, ≥99.9%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'