Learn More
(+/-)-1-Octyn-3-ol, 98%, Thermo Scientific Chemicals
33.90 € - 326.00 €
Chemical Identifiers
CAS | 818-72-4 |
---|---|
Molecular Formula | C8H14O |
Molecular Weight (g/mol) | 126.20 |
MDL Number | MFCD00004588 |
InChI Key | VUGRNZHKYVHZSN-UHFFFAOYNA-N |
Synonym | 1-octyn-3-ol, 1-octyne-3-ol, oct-1-yne-3-ol, 1-oct-1-yn-3-ol, 3-hydroxyoct-1-yne, acmc-20apfv, acmc-20apkf, rac-1-octyn-3-ol, oct-1 yn-3 ol, pubchem11033 |
PubChem CID | 13166 |
IUPAC Name | oct-1-yn-3-ol |
SMILES | CCCCCC(O)C#C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460813
|
Thermo Scientific Alfa Aesar
A13267.09 |
10 g |
33.90 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470813
|
Thermo Scientific Alfa Aesar
A13267.18 |
50 g |
102.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474150
|
Thermo Scientific Alfa Aesar
A13267.30 |
250 g |
326.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
Chemical Identifiers
818-72-4 | |
126.20 | |
VUGRNZHKYVHZSN-UHFFFAOYNA-N | |
13166 | |
CCCCCC(O)C#C |
C8H14O | |
MFCD00004588 | |
1-octyn-3-ol, 1-octyne-3-ol, oct-1-yne-3-ol, 1-oct-1-yn-3-ol, 3-hydroxyoct-1-yne, acmc-20apfv, acmc-20apkf, rac-1-octyn-3-ol, oct-1 yn-3 ol, pubchem11033 | |
oct-1-yn-3-ol |
Specifications
818-72-4 | |
0.865 | |
63°C (145°F) | |
1.442 | |
1098642 | |
Soluble in water. | |
CCCCCC(O)C#C | |
126.20 | |
126.2 | |
(+/-)-1-Octyn-3-ol |
-60°C | |
180°C to 181°C | |
C8H14O | |
MFCD00004588 | |
1-octyn-3-ol, 1-octyne-3-ol, oct-1-yne-3-ol, 1-oct-1-yn-3-ol, 3-hydroxyoct-1-yne, acmc-20apfv, acmc-20apkf, rac-1-octyn-3-ol, oct-1 yn-3 ol, pubchem11033 | |
VUGRNZHKYVHZSN-UHFFFAOYNA-N | |
oct-1-yn-3-ol | |
13166 | |
98% |
Safety and Handling
GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
missing translation for 'einecsNumber' : 212-455-4
missing translation for 'rtecsNumber' : RI2737000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only