missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Octyn-3-ol, 98%, Thermo Scientific Chemicals

Product Code. 11460813
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 818-72-4
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00004588
InChI Key VUGRNZHKYVHZSN-UHFFFAOYNA-N
Synonym 1-octyn-3-ol, 1-octyne-3-ol, oct-1-yne-3-ol, 1-oct-1-yn-3-ol, 3-hydroxyoct-1-yne, acmc-20apfv, acmc-20apkf, rac-1-octyn-3-ol, oct-1 yn-3 ol, pubchem11033
PubChem CID 13166
IUPAC Name oct-1-yn-3-ol
SMILES CCCCCC(O)C#C

Specifications

Melting Point -60°C
Density 0.865
Boiling Point 180°C to 181°C
Flash Point 63°C (145°F)
Refractive Index 1.442
Quantity 10 g
Beilstein 1098642
Solubility Information Soluble in water.
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material (+/-)-1-Octyn-3-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'