missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanol, 98%, Reagent Grade, Honeywell™

Reagent Grade, 98%

19.84 € - 129.04 €

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15677890
View Documents Promotion Details
Honeywell Research Chemicals
H13303-100ML
100 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15687890
View Documents Promotion Details
Honeywell Research Chemicals
H13303-1L
1 L Glass bottle
54.53 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15697890
View Documents Promotion Details
Honeywell Research Chemicals
H13303-2.5L
2.5 L Glass bottle
129.04 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

111-27-3
102.177
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
8103
hexan-1-ol
C6H14O
MFCD00002982
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
CHEBI:87393
CCCCCCO

Specifications

111-27-3
Colorless
156°C to 157°C (literature)
CH3(CH2)5OH
UN2282
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
CCCCCCO
102.177
CHEBI:87393
1mmHg (25.6°C)
1-Hexanol
-52°C (literature)
0.814g/mL at 25°C
C6H14O
MFCD00002982
969167
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
hexan-1-ol
8103
102.17g/mol
0.98
Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338

missing translation for 'dotInformation' : 3/PGIII

missing translation for 'rtecsNumber' : 203-852-3

SDS
Documents

Documents

Product Certifications