missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanol, 98%, Reagent Grade, Honeywell™

Product Code. 15677890
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO

Specifications

Melting Point -52°C (literature)
Color Colorless
Density 0.814g/mL at 25°C
Boiling Point 156°C to 157°C (literature)
Packaging Glass bottle
Linear Formula CH3(CH2)5OH
Quantity 100 mL
UN Number UN2282
Beilstein 969167
Formula Weight 102.17g/mol
Vapor Pressure 1mmHg (25.6°C)
Percent Purity 0.98
Chemical Name or Material 1-Hexanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'