missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Decylboronic acid pinacol ester, 98%, Thermo Scientific™
37.50 € - 542.00 €
Chemical Identifiers
CAS | 141091-38-5 |
---|---|
Molecular Formula | C16H33BO2 |
Molecular Weight (g/mol) | 268.248 |
MDL Number | MFCD12546186 |
InChI Key | HRMCLNRSAPIJDP-UHFFFAOYSA-N |
Synonym | 1-decylboronic acid pinacol ester, 2-n-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
PubChem CID | 11265643 |
IUPAC Name | 2-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)CCCCCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15443927
|
Thermo Scientific Alfa Aesar
H30403.03 |
1 g |
37.50 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15453927
|
Thermo Scientific Alfa Aesar
H30403.06 |
5 g | |||||||
15463927
|
Thermo Scientific Alfa Aesar
H30403.14 |
25 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
141091-38-5 | |
268.248 | |
HRMCLNRSAPIJDP-UHFFFAOYSA-N | |
11265643 | |
B1(OC(C(O1)(C)C)(C)C)CCCCCCCCCC |
C16H33BO2 | |
MFCD12546186 | |
1-decylboronic acid pinacol ester, 2-n-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
2-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
141091-38-5 | |
1.436 | |
1-decylboronic acid pinacol ester, 2-n-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
B1(OC(C(O1)(C)C)(C)C)CCCCCCCCCC | |
268.248 | |
268.25 | |
1-Decylboronic acid pinacol ester |
C16H33BO2 | |
MFCD12546186 | |
HRMCLNRSAPIJDP-UHFFFAOYSA-N | |
2-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
11265643 | |
98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures