missing translation for 'onlineSavingsMsg'
Learn More

1-Decylboronic acid pinacol ester, 98%, Thermo Scientific™

Product Code. 15453927
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141091-38-5
Molecular Formula C16H33BO2
Molecular Weight (g/mol) 268.248
MDL Number MFCD12546186
InChI Key HRMCLNRSAPIJDP-UHFFFAOYSA-N
Synonym 1-decylboronic acid pinacol ester, 2-n-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11265643
IUPAC Name 2-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCCCCCCCCC

Specifications

Refractive Index 1.436
Quantity 5 g
Formula Weight 268.25
Percent Purity 98%
Chemical Name or Material 1-Decylboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'