missing translation for 'onlineSavingsMsg'
Learn More

1-chloro-4-(3-iodopropoxy)benzene, 97%, Thermo Scientific™

16.10 € - 85.30 €

Chemical Identifiers

CAS 119795-57-2
Molecular Formula C9H10ClIO
Molecular Weight (g/mol) 296.532
MDL Number MFCD01764552
InChI Key HVVWUBWHLIPSCS-UHFFFAOYSA-N
Synonym 1-chloro-4-3-iodopropoxy benzene, 3-4-chlorophenoxy propyl iodide, benzene,1-chloro-4-3-iodopropoxy, acmc-1c0gn, 1-4-chlorophenoxy-3-iodopropane
PubChem CID 2794890
IUPAC Name 1-chloro-4-(3-iodopropoxy)benzene
SMILES C1=CC(=CC=C1OCCCI)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10099321
View Documents
Thermo Scientific Maybridge
RH01447DA
1 g Amber Glass Bottle
16.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
10039301
View Documents
Thermo Scientific Maybridge
RH01447EA
10 g Amber Glass Bottle
85.30 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

119795-57-2
296.532
HVVWUBWHLIPSCS-UHFFFAOYSA-N
2794890
C1=CC(=CC=C1OCCCI)Cl
C9H10ClIO
MFCD01764552
1-chloro-4-3-iodopropoxy benzene, 3-4-chlorophenoxy propyl iodide, benzene,1-chloro-4-3-iodopropoxy, acmc-1c0gn, 1-4-chlorophenoxy-3-iodopropane
1-chloro-4-(3-iodopropoxy)benzene

Specifications

119795-57-2
97%
ClC6H4OCH2CH2CH2I
1-chloro-4-3-iodopropoxy benzene, 3-4-chlorophenoxy propyl iodide, benzene,1-chloro-4-3-iodopropoxy, acmc-1c0gn, 1-4-chlorophenoxy-3-iodopropane
C1=CC(=CC=C1OCCCI)Cl
296.532
296.53
1-chloro-4-(3-iodopropoxy)benzene
97.0
C9H10ClIO
MFCD01764552
HVVWUBWHLIPSCS-UHFFFAOYSA-N
1-chloro-4-(3-iodopropoxy)benzene
2794890
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.