missing translation for 'onlineSavingsMsg'
Learn More

1-chloro-4-(3-iodopropoxy)benzene, 97%, Thermo Scientific™

Product Code. 10039301
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 119795-57-2
Molecular Formula C9H10ClIO
Molecular Weight (g/mol) 296.532
MDL Number MFCD01764552
InChI Key HVVWUBWHLIPSCS-UHFFFAOYSA-N
Synonym 1-chloro-4-3-iodopropoxy benzene, 3-4-chlorophenoxy propyl iodide, benzene,1-chloro-4-3-iodopropoxy, acmc-1c0gn, 1-4-chlorophenoxy-3-iodopropane
PubChem CID 2794890
IUPAC Name 1-chloro-4-(3-iodopropoxy)benzene
SMILES C1=CC(=CC=C1OCCCI)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula ClC6H4OCH2CH2CH2I
Quantity 10 g
Formula Weight 296.53
Percent Purity 97%
Chemical Name or Material 1-chloro-4-(3-iodopropoxy)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.