missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminomethyl-1-cyclohexanol hydrochloride, 98%, Thermo Scientific Chemicals
30.54 € - 97.45 €
Chemical Identifiers
CAS | 19968-85-5 |
---|---|
Molecular Formula | C7H15NO·HCl |
Molecular Weight (g/mol) | 165.67 |
MDL Number | MFCD00003859 |
InChI Key | DXIAEURKHWGAFH-UHFFFAOYSA-N |
Synonym | 1-aminomethyl-1-cyclohexanol hydrochloride, 1-aminomethyl cyclohexanol hydrochloride, 1-aminomethylcyclohexan-1-ol hydrochloride, 1-aminomethyl-cyclohexanol hydrochloride, 1-aminomethyl cyclohexan-1-ol hydrochloride, 1-aminomethylcyclohexan-1-ol hcl, acmc-20aoy8, 1-aminomethyl-1-cyclohexanol hcl, 1-aminomethylcyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrogen chloride |
PubChem CID | 3084332 |
IUPAC Name | 1-(aminomethyl)cyclohexan-1-ol;hydrochloride |
SMILES | C1CCC(CC1)(CN)O.Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19968-85-5 | |
165.67 | |
DXIAEURKHWGAFH-UHFFFAOYSA-N | |
3084332 | |
C1CCC(CC1)(CN)O.Cl |
C7H15NO·HCl | |
MFCD00003859 | |
1-aminomethyl-1-cyclohexanol hydrochloride, 1-aminomethyl cyclohexanol hydrochloride, 1-aminomethylcyclohexan-1-ol hydrochloride, 1-aminomethyl-cyclohexanol hydrochloride, 1-aminomethyl cyclohexan-1-ol hydrochloride, 1-aminomethylcyclohexan-1-ol hcl, acmc-20aoy8, 1-aminomethyl-1-cyclohexanol hcl, 1-aminomethylcyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrogen chloride | |
1-(aminomethyl)cyclohexan-1-ol;hydrochloride |
Specifications
19968-85-5 | |
100.0 | |
Beige-Gray to White | |
97.5% min. (Argentometry) | |
H2NCH2C6H10OH·HCl | |
1-aminomethyl-1-cyclohexanol hydrochloride, 1-aminomethyl cyclohexanol hydrochloride, 1-aminomethylcyclohexan-1-ol hydrochloride, 1-aminomethyl-cyclohexanol hydrochloride, 1-aminomethyl cyclohexan-1-ol hydrochloride, 1-aminomethylcyclohexan-1-ol hcl, acmc-20aoy8, 1-aminomethyl-1-cyclohexanol hcl, 1-aminomethylcyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrogen chloride | |
C1CCC(CC1)(CN)O.Cl | |
165.67 | |
165.67 | |
Powder |
97.5 | |
217°C to 219°C | |
Authentic | |
C7H15NO·HCl | |
MFCD00003859 | |
DXIAEURKHWGAFH-UHFFFAOYSA-N | |
1-(aminomethyl)cyclohexan-1-ol;hydrochloride | |
3084332 | |
98% | |
1-Aminomethyl-1-cyclohexanol hydrochloride, 98% |
Safety and Handling
missing translation for 'einecsNumber' : 243-444-2
missing translation for 'tsca' : TSCA
Product Identifier
- 1-Aminomethyl-1-cyclohexanol hydrochloride
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title