missing translation for 'onlineSavingsMsg'
Learn More

1-Aminomethyl-1-cyclohexanol hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10459670
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19968-85-5
Molecular Formula C7H15NO·HCl
Molecular Weight (g/mol) 165.67
MDL Number MFCD00003859
InChI Key DXIAEURKHWGAFH-UHFFFAOYSA-N
Synonym 1-aminomethyl-1-cyclohexanol hydrochloride, 1-aminomethyl cyclohexanol hydrochloride, 1-aminomethylcyclohexan-1-ol hydrochloride, 1-aminomethyl-cyclohexanol hydrochloride, 1-aminomethyl cyclohexan-1-ol hydrochloride, 1-aminomethylcyclohexan-1-ol hcl, acmc-20aoy8, 1-aminomethyl-1-cyclohexanol hcl, 1-aminomethylcyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrogen chloride
PubChem CID 3084332
IUPAC Name 1-(aminomethyl)cyclohexan-1-ol;hydrochloride
SMILES C1CCC(CC1)(CN)O.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 217°C to 219°C
Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NCH2C6H10OH·HCl
Quantity 5 g
Formula Weight 165.67
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Aminomethyl-1-cyclohexanol hydrochloride, 98%
Show More Show Less
Product Identifier
  • 1-Aminomethyl-1-cyclohexanol hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.