missing translation for 'onlineSavingsMsg'
Learn More

1,5,9-Triazacyclotridecane trihydrobromide, 98%, Thermo Scientific™

53.00 € - 208.00 €

Chemical Identifiers

CAS 74676-52-1
Molecular Formula C10H26Br3N3
Molecular Weight (g/mol) 428.051
MDL Number MFCD00013326
InChI Key HRDCXXGINBVBMF-UHFFFAOYSA-N
Synonym 1,5,9-triazacyclotridecane trihydrobromide, 1,5,9-triazacyclotridecane-hydrogen bromide 1/3
PubChem CID 24212082
IUPAC Name 1,5,9-triazacyclotridecane;trihydrobromide
SMILES C1CCNCCCNCCCNC1.Br.Br.Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478487
View Documents
Thermo Scientific Alfa Aesar
A18560.MC
100 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11488487
View Documents
Thermo Scientific Alfa Aesar
A18560.ME
500 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

74676-52-1
428.051
HRDCXXGINBVBMF-UHFFFAOYSA-N
24212082
C1CCNCCCNCCCNC1.Br.Br.Br
C10H26Br3N3
MFCD00013326
1,5,9-triazacyclotridecane trihydrobromide, 1,5,9-triazacyclotridecane-hydrogen bromide 1/3
1,5,9-triazacyclotridecane;trihydrobromide

Specifications

74676-52-1
C10H26Br3N3
Hygroscopic
HRDCXXGINBVBMF-UHFFFAOYSA-N
1,5,9-triazacyclotridecane;trihydrobromide
24212082
98%
∼300°C (decomposition)
MFCD00013326
1,5,9-triazacyclotridecane trihydrobromide, 1,5,9-triazacyclotridecane-hydrogen bromide 1/3
C1CCNCCCNCCCNC1.Br.Br.Br
428.051
428.05
1,5,9-Triazacyclotridecane trihydrobromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.