missing translation for 'onlineSavingsMsg'
Learn More

1,5,9-Triazacyclotridecane trihydrobromide, 98%, Thermo Scientific™

Product Code. 11488487
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74676-52-1
Molecular Formula C10H26Br3N3
Molecular Weight (g/mol) 428.051
MDL Number MFCD00013326
InChI Key HRDCXXGINBVBMF-UHFFFAOYSA-N
Synonym 1,5,9-triazacyclotridecane trihydrobromide, 1,5,9-triazacyclotridecane-hydrogen bromide 1/3
PubChem CID 24212082
IUPAC Name 1,5,9-triazacyclotridecane;trihydrobromide
SMILES C1CCNCCCNCCCNC1.Br.Br.Br

Specifications

Melting Point ∼300°C (decomposition)
Quantity 500 mg
Sensitivity Hygroscopic
Formula Weight 428.05
Percent Purity 98%
Chemical Name or Material 1,5,9-Triazacyclotridecane trihydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.