Learn More
1-(4-Methylphenyl)ethanol, 97%, Thermo Scientific Chemicals
27.00 € - 344.00 €
Chemical Identifiers
CAS | 536-50-5 |
---|---|
Molecular Formula | C9H12O |
Molecular Weight (g/mol) | 136.194 |
MDL Number | MFCD00016860 |
InChI Key | JESIHYIJKKUWIS-UHFFFAOYSA-N |
Synonym | 1-4-methylphenyl ethanol, 1-p-tolyl ethanol, bilagen, galinex, norbilan, tomobil, methyl-p-tolylcarbinol, p-tolylmethylcarbinol, 1-4-tolyl ethanol, 1-p-tolyl-1-ethanol |
PubChem CID | 10817 |
IUPAC Name | 1-(4-methylphenyl)ethanol |
SMILES | CC1=CC=C(C=C1)C(C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11476390
|
Thermo Scientific Alfa Aesar
B20832.03 |
1 g |
27.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11486390
|
Thermo Scientific Alfa Aesar
B20832.06 |
5 g |
93.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11496390
|
Thermo Scientific Alfa Aesar
B20832.14 |
25 g |
344.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.
Solubility
Slightly miscible with water.
Notes
Incompatible with oxidizing agents and strong reducing agents. Store in a cool place.
Chemical Identifiers
536-50-5 | |
136.194 | |
JESIHYIJKKUWIS-UHFFFAOYSA-N | |
10817 | |
CC1=CC=C(C=C1)C(C)O |
C9H12O | |
MFCD00016860 | |
1-4-methylphenyl ethanol, 1-p-tolyl ethanol, bilagen, galinex, norbilan, tomobil, methyl-p-tolylcarbinol, p-tolylmethylcarbinol, 1-4-tolyl ethanol, 1-p-tolyl-1-ethanol | |
1-(4-methylphenyl)ethanol |
Specifications
536-50-5 | |
218°C to 220°C | |
C9H12O | |
MFCD00016860 | |
14,3237 | |
Slightly miscible with water. | |
CC1=CC=C(C=C1)C(C)O | |
136.194 | |
136.2 | |
1-(4-Methylphenyl)ethanol |
0.987 | |
107°C (225°F) | |
1.523 | |
1857562 | |
1-4-methylphenyl ethanol, 1-p-tolyl ethanol, bilagen, galinex, norbilan, tomobil, methyl-p-tolylcarbinol, p-tolylmethylcarbinol, 1-4-tolyl ethanol, 1-p-tolyl-1-ethanol | |
JESIHYIJKKUWIS-UHFFFAOYSA-N | |
1-(4-methylphenyl)ethanol | |
10817 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 208-637-8
missing translation for 'rtecsNumber' : DO4565000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only