missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methylphenyl)ethanol, 97%, Thermo Scientific Chemicals

Product Code. 11486390
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents and strong reducing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-50-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00016860
InChI Key JESIHYIJKKUWIS-UHFFFAOYSA-N
Synonym 1-4-methylphenyl ethanol, 1-p-tolyl ethanol, bilagen, galinex, norbilan, tomobil, methyl-p-tolylcarbinol, p-tolylmethylcarbinol, 1-4-tolyl ethanol, 1-p-tolyl-1-ethanol
PubChem CID 10817
IUPAC Name 1-(4-methylphenyl)ethanol
SMILES CC1=CC=C(C=C1)C(C)O

Specifications

Density 0.987
Boiling Point 218°C to 220°C
Flash Point 107°C (225°F)
Refractive Index 1.523
Quantity 5 g
Beilstein 1857562
Merck Index 14,3237
Solubility Information Slightly miscible with water.
Formula Weight 136.2
Percent Purity 97%
Chemical Name or Material 1-(4-Methylphenyl)ethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'