missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Amino-2-hydroxyphenyl)ethan-1-one, Tech., Thermo Scientific™
58.00 € - 290.00 €
Chemical Identifiers
CAS | 2476-29-1 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.17 |
MDL Number | MFCD00100636 |
InChI Key | QQZFVONVJPXCSQ-UHFFFAOYSA-N |
Synonym | 1-4-amino-2-hydroxyphenyl ethanone, 1-4-amino-2-hydroxyphenyl ethan-1-one, 4'-amino-2'-hydroxyacetophenone, 1-4-amino-2-hydroxyphenyl ethane-1-one, ethanone, 1-4-amino-2-hydroxyphenyl, 1-4-amino-2-hydroxy-phenyl ethanone, 2-acetyl-5-aminophenol, intermediates-zcf02667, amino-2-hydroxyphenyl ethanone, 4-acetyl-3-hydroxyaniline |
PubChem CID | 459296 |
IUPAC Name | 1-(4-amino-2-hydroxyphenyl)ethanone |
SMILES | CC(=O)C1=C(O)C=C(N)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10748304
|
Thermo Scientific Maybridge
BTB10056DA |
1 g | Amber Glass Bottle |
58.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10644903
|
Thermo Scientific Maybridge
BTB10056EA |
10 g | Amber Glass Bottle |
290.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
2476-29-1 | |
151.17 | |
QQZFVONVJPXCSQ-UHFFFAOYSA-N | |
459296 | |
CC(=O)C1=C(O)C=C(N)C=C1 |
C8H9NO2 | |
MFCD00100636 | |
1-4-amino-2-hydroxyphenyl ethanone, 1-4-amino-2-hydroxyphenyl ethan-1-one, 4'-amino-2'-hydroxyacetophenone, 1-4-amino-2-hydroxyphenyl ethane-1-one, ethanone, 1-4-amino-2-hydroxyphenyl, 1-4-amino-2-hydroxy-phenyl ethanone, 2-acetyl-5-aminophenol, intermediates-zcf02667, amino-2-hydroxyphenyl ethanone, 4-acetyl-3-hydroxyaniline | |
1-(4-amino-2-hydroxyphenyl)ethanone |
Specifications
2476-29-1 | |
C8H9NO2 | |
1-4-amino-2-hydroxyphenyl ethanone, 1-4-amino-2-hydroxyphenyl ethan-1-one, 4'-amino-2'-hydroxyacetophenone, 1-4-amino-2-hydroxyphenyl ethane-1-one, ethanone, 1-4-amino-2-hydroxyphenyl, 1-4-amino-2-hydroxy-phenyl ethanone, 2-acetyl-5-aminophenol, intermediates-zcf02667, amino-2-hydroxyphenyl ethanone, 4-acetyl-3-hydroxyaniline | |
CC(=O)C1=C(O)C=C(N)C=C1 | |
151.17 | |
151.16 | |
Technical Grade |
97% | |
MFCD00100636 | |
QQZFVONVJPXCSQ-UHFFFAOYSA-N | |
1-(4-amino-2-hydroxyphenyl)ethanone | |
459296 | |
Tech. | |
1-(4-Amino-2-hydroxyphenyl)ethan-1-one |
Safety and Handling
Warning