missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Amino-2-hydroxyphenyl)ethan-1-one, Tech., Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 151.16 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 1-(4-Amino-2-hydroxyphenyl)ethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'