missing translation for 'onlineSavingsMsg'
Learn More

(1,2-Dibromoethyl)benzene, 97%, Thermo Scientific Chemicals

25.20 € - 170.00 €

Chemical Identifiers

CAS 93-52-7
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000138
InChI Key SHKKTLSDGJRCTR-UHFFFAOYSA-N
Synonym 1,2-dibromoethyl benzene, dowspray 9, 1,2-dibromo-1-phenylethane, 1,2-dibromo phenyl ethane, benzene, 1,2-dibromoethyl, 1,2-dibromo-2-phenylethane, vic-styrene dibromide, alpha,beta-dibromoethylbenzene, ccris 2090, .alpha.,.beta.-dibromoethylbenzene
PubChem CID 7145
IUPAC Name 1,2-dibromoethylbenzene
SMILES C1=CC=C(C=C1)C(CBr)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11491567
View Documents
Thermo Scientific Alfa Aesar
B24990.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11401577
View Documents
Thermo Scientific Alfa Aesar
B24990.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11411577
View Documents
Thermo Scientific Alfa Aesar
B24990.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

93-52-7
263.96
SHKKTLSDGJRCTR-UHFFFAOYSA-N
7145
C1=CC=C(C=C1)C(CBr)Br
C8H8Br2
MFCD00000138
1,2-dibromoethyl benzene, dowspray 9, 1,2-dibromo-1-phenylethane, 1,2-dibromo phenyl ethane, benzene, 1,2-dibromoethyl, 1,2-dibromo-2-phenylethane, vic-styrene dibromide, alpha,beta-dibromoethylbenzene, ccris 2090, .alpha.,.beta.-dibromoethylbenzene
1,2-dibromoethylbenzene

Specifications

93-52-7
139°C to 140°C (15 mmHg)
MFCD00000138
1,2-dibromoethyl benzene, dowspray 9, 1,2-dibromo-1-phenylethane, 1,2-dibromo phenyl ethane, benzene, 1,2-dibromoethyl, 1,2-dibromo-2-phenylethane, vic-styrene dibromide, alpha,beta-dibromoethylbenzene, ccris 2090, .alpha.,.beta.-dibromoethylbenzene
C1=CC=C(C=C1)C(CBr)Br
263.96
263.96
(1,2-Dibromoethyl)benzene
70°C to 74°C
C8H8Br2
UN3261
SHKKTLSDGJRCTR-UHFFFAOYSA-N
1,2-dibromoethylbenzene
7145
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 202-253-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only