missing translation for 'onlineSavingsMsg'
Learn More

(1,2-Dibromoethyl)benzene, 97%, Thermo Scientific Chemicals

Product Code. 11401577
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-52-7
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000138
InChI Key SHKKTLSDGJRCTR-UHFFFAOYSA-N
Synonym 1,2-dibromoethyl benzene, dowspray 9, 1,2-dibromo-1-phenylethane, 1,2-dibromo phenyl ethane, benzene, 1,2-dibromoethyl, 1,2-dibromo-2-phenylethane, vic-styrene dibromide, alpha,beta-dibromoethylbenzene, ccris 2090, .alpha.,.beta.-dibromoethylbenzene
PubChem CID 7145
IUPAC Name 1,2-dibromoethylbenzene
SMILES C1=CC=C(C=C1)C(CBr)Br

Specifications

Melting Point 70°C to 74°C
Boiling Point 139°C to 140°C (15 mmHg)
Quantity 100 g
UN Number UN3261
Formula Weight 263.96
Percent Purity 97%
Chemical Name or Material (1,2-Dibromoethyl)benzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'