Learn More
1,2-Cyclohexanedione, 98+%, Thermo Scientific Chemicals
66.70 € - 728.00 €
Chemical Identifiers
CAS | 765-87-7 |
---|---|
Molecular Formula | C6H8O2 |
MDL Number | MFCD00001648 |
InChI Key | OILAIQUEIWYQPH-UHFFFAOYSA-N |
Synonym | 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol |
PubChem CID | 13006 |
ChEBI | CHEBI:41674 |
IUPAC Name | cyclohexane-1,2-dione |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11407017
|
Thermo Scientific Alfa Aesar
A14401.06 |
5 g |
66.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417017
|
Thermo Scientific Alfa Aesar
A14401.14 |
25 g |
208.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427017
|
Thermo Scientific Alfa Aesar
A14401.22 |
100 g |
728.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,2-Cyclohexanedione is utilized as a substrate to study enzyme cyclohexane-1,2-dione hydrolase, which is a new tool to degrade alicyclic compounds. It also acts as a specific reagent for arginine residues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Cyclohexanedione is utilized as a substrate to study enzyme cyclohexane-1,2-dione hydrolase, which is a new tool to degrade alicyclic compounds. It also acts as a specific reagent for arginine residues.
Solubility
Soluble in water and alcohol.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
765-87-7 | |
MFCD00001648 | |
1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol | |
CHEBI:41674 |
C6H8O2 | |
OILAIQUEIWYQPH-UHFFFAOYSA-N | |
13006 | |
cyclohexane-1,2-dione |
Specifications
765-87-7 | |
1.12 | |
84°C (183°F) | |
1.519 | |
507419 | |
Soluble in water and alcohol. | |
C1CCC(=O)C(=O)C1 | |
112.128 | |
CHEBI:41674 | |
≥98% |
35°C to 38°C | |
193°C to 195°C | |
C6H8O2 | |
MFCD00001648 | |
1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol | |
OILAIQUEIWYQPH-UHFFFAOYSA-N | |
cyclohexane-1,2-dione | |
13006 | |
112.13 | |
1,2-Cyclohexanedione |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 212-155-3
missing translation for 'rtecsNumber' : GV0340000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only