missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanedione, 98+%, Thermo Scientific Chemicals

Product Code. 11427017
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,2-Cyclohexanedione is utilized as a substrate to study enzyme cyclohexane-1,2-dione hydrolase, which is a new tool to degrade alicyclic compounds. It also acts as a specific reagent for arginine residues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Cyclohexanedione is utilized as a substrate to study enzyme cyclohexane-1,2-dione hydrolase, which is a new tool to degrade alicyclic compounds. It also acts as a specific reagent for arginine residues.

Solubility
Soluble in water and alcohol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-87-7
Molecular Formula C6H8O2
MDL Number MFCD00001648
InChI Key OILAIQUEIWYQPH-UHFFFAOYSA-N
Synonym 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol
PubChem CID 13006
ChEBI CHEBI:41674
IUPAC Name cyclohexane-1,2-dione

Specifications

Melting Point 35°C to 38°C
Density 1.12
Boiling Point 193°C to 195°C
Flash Point 84°C
Refractive Index 1.519
Quantity 100 g
Beilstein 507419
Solubility Information Soluble aq. soln. and alcohol.
SMILES O=C1CCCCC1=O
Molecular Weight (g/mol) 112.13
Formula Weight 112.13
Percent Purity ≥98%
Chemical Name or Material 1,2-Cyclohexanedione
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'