Learn More
1,12-Dodecanediol, 99%, Thermo Scientific Chemicals
44.10 € - 122.00 €
Chemical Identifiers
CAS | 5675-51-4 |
---|---|
Molecular Formula | C12H26O2 |
Molecular Weight (g/mol) | 202.34 |
MDL Number | MFCD00004755 |
InChI Key | GHLKSLMMWAKNBM-UHFFFAOYSA-N |
Synonym | 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h |
PubChem CID | 79758 |
IUPAC Name | dodecane-1,12-diol |
SMILES | OCCCCCCCCCCCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379789
|
Thermo Scientific Alfa Aesar
B22885.14 |
25 g |
44.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389789
|
Thermo Scientific Alfa Aesar
B22885.22 |
100 g |
122.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.
Solubility
Soluble in alcohol and warm ether. Insoluble in water and petroleum ether.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
5675-51-4 | |
202.34 | |
GHLKSLMMWAKNBM-UHFFFAOYSA-N | |
79758 | |
OCCCCCCCCCCCCO |
C12H26O2 | |
MFCD00004755 | |
1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h | |
dodecane-1,12-diol |
Specifications
5675-51-4 | |
188°C to 190°C (12 mmHg) | |
MFCD00004755 | |
1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h | |
GHLKSLMMWAKNBM-UHFFFAOYSA-N | |
dodecane-1,12-diol | |
79758 | |
99% |
81°C to 84°C | |
C12H26O2 | |
1742760 | |
Soluble in alcohol and warm ether. Insoluble in water and petroleum ether. | |
OCCCCCCCCCCCCO | |
202.34 | |
202.34 | |
1,12-Dodecanediol |
Safety and Handling
missing translation for 'einecsNumber' : 227-133-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only