missing translation for 'onlineSavingsMsg'
Learn More

1,12-Dodecanediol, 99%, Thermo Scientific Chemicals

Product Code. 11389789
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.

Solubility
Soluble in alcohol and warm ether. Insoluble in water and petroleum ether.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5675-51-4
Molecular Formula C12H26O2
Molecular Weight (g/mol) 202.34
MDL Number MFCD00004755
InChI Key GHLKSLMMWAKNBM-UHFFFAOYSA-N
Synonym 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h
PubChem CID 79758
IUPAC Name dodecane-1,12-diol
SMILES OCCCCCCCCCCCCO

Specifications

Melting Point 81°C to 84°C
Boiling Point 188°C to 190°C (12 mmHg)
Quantity 100 g
Beilstein 1742760
Solubility Information Soluble in alcohol and warm ether. Insoluble in water and petroleum ether.
Formula Weight 202.34
Percent Purity 99%
Chemical Name or Material 1,12-Dodecanediol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'