missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,2-Tetrachloroethane, Reagent grade, ≥98.0%, Honeywell™
Reagent Grade,≥98.0%
49.61 € - 173.59 €
Chemical Identifiers
CAS | 79-34-5 |
---|---|
Molecular Formula | C2H2Cl4 |
Molecular Weight (g/mol) | 167.838 |
MDL Number | MFCD00000848 |
InChI Key | QPFMBZIOSGYJDE-UHFFFAOYSA-N |
Synonym | s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan |
PubChem CID | 6591 |
ChEBI | CHEBI:36026 |
IUPAC Name | 1,1,2,2-tetrachloroethane |
SMILES | C(C(Cl)Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15661670
|
Honeywell Research Chemicals
185434-100ML |
100 mL | Glass Bottle | ||||||
15691670
|
Honeywell Research Chemicals
185434-500ML |
500 mL | Glass Bottle | ||||||
15671670
|
Honeywell Research Chemicals
185434-1L |
1 L | Glass Bottle | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
79-34-5 | |
167.838 | |
QPFMBZIOSGYJDE-UHFFFAOYSA-N | |
6591 | |
1,1,2,2-tetrachloroethane |
C2H2Cl4 | |
MFCD00000848 | |
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan | |
CHEBI:36026 | |
C(C(Cl)Cl)(Cl)Cl |
Specifications
79-34-5 | |
Colorless | |
147°C | |
CHCl2CHCl2 | |
MFCD00000848 | |
969206 | |
QPFMBZIOSGYJDE-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethane | |
6591 | |
CHEBI:36026 | |
8mmHg (20°C) | |
Reagent | |
1,1,2,2-Tetrachloroethane |
-43°C | |
1.586g/mL at 25°C | |
C2H2Cl4 | |
n20/D 1.494(lit.) | |
UN1702 | |
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan | |
C(C(Cl)Cl)(Cl)Cl | |
167.838 | |
5.8 (vs air) | |
167.85g/mol | |
≥98.0% | |
Liquid |
Safety and Handling
P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310-P361 + P364
H310 + H330-H411
missing translation for 'dotInformation' : 6.1/PGII
missing translation for 'einecsNumber' : 201-197-8
missing translation for 'rtecsNumber' : KI8575000