missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,2-Tetrachloroethane, Reagent grade, ≥98.0%, Honeywell™
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | -43°C |
Color | Colorless |
Density | 1.586g/mL at 25°C |
Boiling Point | 147°C |
Packaging | Glass Bottle |
Linear Formula | CHCl2CHCl2 |
Refractive Index | n20/D 1.494(lit.) |
Quantity | 1 L |
UN Number | UN1702 |
Beilstein | 969206 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'