missing translation for 'onlineSavingsMsg'
Learn More

ZNF259 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen ZNF259
Concentration 0.45 mg/mL
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16834794
View Documents
Proteintech
15558-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16824794
View Documents
Proteintech
15558-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MW of this protein is 51 kDa, and this antibody specifically recognizes the 51 kDa protein.

ZNF259, also named as Zpr1, is an evolutionarily conserved protein characterized by two C4 zinc fingers and two Zpr1-specific conserved homology domains. Zpr1 was first identified in mammalian cells as a cytoplasmic protein that was capable of binding the intracellular domain of the inactivated EGF receptor (EGFR).
Specifications

Specifications

ZNF259
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
O75312
8882
ZNF259 Fusion Protein Ag7980
Primary
-20°C
ZPR1
0.45 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
zinc finger protein 259, Zinc finger protein ZPR1, ZNF259, ZPR1
ZPR1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.