Learn More
ZNF143 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16618-1-AP-20UL
 View more versions of this product
Product Code. 16816294
  • 145.00 € / 20µL
Explore more special offers
Description
Description
Transcriptional activator. Activates the gene for selenocysteine tRNA (tRNAsec). Binds to the SPH motif of small nuclear RNA (snRNA) gene promoters. Participates in efficient U6 RNA polymerase III transcription via its interaction with CHD8.
Specifications
Specifications
ZNF143
Polyclonal
Unconjugated
ZNF143
hStaf, pHZ 1, SBF, SPH binding factor, STAF, zinc finger protein 143, ZNF143
Rabbit
Antigen Affinity Chromatography
RUO
20841, 361627, 7702
-20°C
Liquid
Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70230, P52747, Q5XIU2
Znf143
ZNF143 Fusion Protein Ag9914
20 μL
Primary
Rat, Mouse, Human