missing translation for 'onlineSavingsMsg'
Learn More

Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf, Thermo Scientific Chemicals

77.08 € - 246.23 €

Chemical Identifiers

CAS 14985-18-3
Molecular Formula N2O7Zr
Molecular Weight (g/mol) 231.23
MDL Number MFCD00149896
InChI Key JWBWFTANBHYHBC-UHFFFAOYSA-N
Synonym zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis
PubChem CID 16211674
IUPAC Name oxozirconium(2+);dinitrate;hydrate
SMILES [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10573771
SDS View Documents
Thermo Scientific Acros
222631000
100 g Glass bottle
77.08 €
100g
Please to purchase this item. Need a web account? Register with us today!
10174750
SDS View Documents
Thermo Scientific Acros
222635000
500 g Glass bottle
246.23 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.