missing translation for 'onlineSavingsMsg'
Learn More

Zirconium wire, 0.25mm (0.01in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

100.50 € - 311.55 €

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11356547
SDS View Documents
Thermo Scientific Alfa Aesar
000416.BW
50 cm
100.50 €
50cm
Please to purchase this item. Need a web account? Register with us today!
11366547
SDS View Documents
Thermo Scientific Alfa Aesar
000416.CB
250 cm
311.55 €
250cm
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-67-7
91.22
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303 MFCD00148878
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]

Specifications

7440-67-7
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
23995
99.95%
≈0.32g/m
(metals basis)
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
[Zr]
91.22
CHEBI:33342
Odorless
Wire
Zirconium wire, 0.25mm (0.01 in.) dia.
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-176-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium wire, 0.25mm (0.01in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.