missing translation for 'onlineSavingsMsg'
Learn More

Zirconium oxide, Yttria stabilized, Grinding Pieces; Diameter (mm), 5, Thermo Scientific Chemicals

Product Code. 11389408
Click to view available options
Quantity:
250 g
1 kg

Specifications: Yttrium oxide (mole) 3%, Hardness 12, Total impuroty level <0.1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00145546

Specifications

Color White
Quantity 1 kg
Odor Odorless
Appearance Off-white chunks
Density 6.02
Chemical Name or Material Zirconium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.