missing translation for 'onlineSavingsMsg'
Learn More

Zirconium oxide, Yttria stabilized, Grinding Pieces; Diameter (mm), 10, Thermo Scientific Chemicals

133.67 € - 471.35 €

Chemical Identifiers

MDL Number MFCD00145546
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11399408
SDS View Documents
Thermo Scientific Alfa Aesar
043286.30
250 g
133.67 €
250g
Please to purchase this item. Need a web account? Register with us today!
11309418
SDS View Documents
Thermo Scientific Alfa Aesar
043286.A1
1 kg
471.35 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00145546

Specifications

White
Odorless
Zirconium oxide
MFCD00145546
Off-white chunks
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-227-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium oxide, Yttria stabilized, Grinding Pieces; Diameter (mm), 10, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.