Learn More
Zirconium(IV) sulfate tetrahydrate, 98+% (metals basis), Thermo Scientific Chemicals
Catalyst support
44.70 € - 432.00 €
Chemical Identifiers
CAS | 7446-31-3 |
---|---|
Molecular Formula | H8O12S2Zr |
Molecular Weight (g/mol) | 355.40 |
MDL Number | MFCD00149267 |
InChI Key | NLOQZPHAWQDLQW-UHFFFAOYSA-J |
Synonym | zirconium sulfate tetrahydrate, unii-os8l03qu95, sulfuric acid, zirconium 4+ salt 2:1 , tetrahydrate, zirconium iv sulfate tetrahydrate, zirconiumsulfatetetrahydrate, 2so4.zr.4h2o, ksc498a6d, zirconium 4+ tetrahydrate disulfate, zirconium 4+ ion tetrahydrate disulfate |
PubChem CID | 23219663 |
IUPAC Name | zirconium(4+);disulfate;tetrahydrate |
SMILES | O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369457
|
Thermo Scientific Alfa Aesar
041041.22 |
100 g |
44.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349457
|
Thermo Scientific Alfa Aesar
041041.36 |
500 g |
136.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359457
|
Thermo Scientific Alfa Aesar
041041.A3 |
2 kg |
432.00 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catalyst support.Zirconium disulfate tetrahydrate is used in organic synthesis. It is also used in tanning white leather. Further, it is used as catalyst supports to precipitate proteins and amino acids. It is also useful in chemical analysis. It plays an essential role as an additive for lubricants and as a pigment stabilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7446-31-3 | |
355.40 | |
NLOQZPHAWQDLQW-UHFFFAOYSA-J | |
23219663 | |
O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
H8O12S2Zr | |
MFCD00149267 | |
zirconium sulfate tetrahydrate, unii-os8l03qu95, sulfuric acid, zirconium 4+ salt 2:1 , tetrahydrate, zirconium iv sulfate tetrahydrate, zirconiumsulfatetetrahydrate, 2so4.zr.4h2o, ksc498a6d, zirconium 4+ tetrahydrate disulfate, zirconium 4+ ion tetrahydrate disulfate | |
zirconium(4+);disulfate;tetrahydrate |
Specifications
7446-31-3 | |
H8O12S2Zr | |
14,10182 | |
Soluble in water. | |
O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
355.40 | |
355.40 (283.34 Anhydrous) | |
3.22 g/mL | |
Zirconium(IV) sulfate tetrahydrate |
White | |
MFCD00149267 | |
zirconium sulfate tetrahydrate, unii-os8l03qu95, sulfuric acid, zirconium 4+ salt 2:1 , tetrahydrate, zirconium iv sulfate tetrahydrate, zirconiumsulfatetetrahydrate, 2so4.zr.4h2o, ksc498a6d, zirconium 4+ tetrahydrate disulfate, zirconium 4+ ion tetrahydrate disulfate | |
NLOQZPHAWQDLQW-UHFFFAOYSA-J | |
zirconium(4+);disulfate;tetrahydrate | |
23219663 | |
≥98% | |
(metals basis) |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 238-694-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only