Learn More
Zirconium aluminide, 99% (metals basis), Thermo Scientific™
161.00 € - 491.00 €
Chemical Identifiers
CAS | 12004-83-0 |
---|---|
Molecular Formula | Al3Zr |
Molecular Weight (g/mol) | 172.169 |
MDL Number | MFCD00064837 |
InChI Key | PGLXYLYDLCVKEE-UHFFFAOYSA-N |
Synonym | zirconium aluminum, aluminum, compd. with zirconium 3:1, zirconium aluminide, trialumane zirconium |
PubChem CID | 6336544 |
IUPAC Name | aluminum;zirconium |
SMILES | [Al].[Al].[Al].[Zr] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11308117
|
Thermo Scientific Alfa Aesar
022886.14 |
25 g |
161.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398107
|
Thermo Scientific Alfa Aesar
022886.22 |
100 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
12004-83-0 | |
172.169 | |
PGLXYLYDLCVKEE-UHFFFAOYSA-N | |
6336544 | |
[Al].[Al].[Al].[Zr] |
Al3Zr | |
MFCD00064837 | |
zirconium aluminum, aluminum, compd. with zirconium 3:1, zirconium aluminide, trialumane zirconium | |
aluminum;zirconium |
Specifications
12004-83-0 | |
Al3Zr | |
zirconium aluminum, aluminum, compd. with zirconium 3:1, zirconium aluminide, trialumane zirconium | |
PGLXYLYDLCVKEE-UHFFFAOYSA-N | |
aluminum;zirconium | |
6336544 | |
99% | |
Zirconium aluminide |
Lump | |
MFCD00064837 | |
Insoluble in water. | |
[Al].[Al].[Al].[Zr] | |
172.169 | |
172.17 | |
(metals basis) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 234-462-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Zirconium aluminide
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.