missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, 0001 Orientation, Thermo Scientific™
Click to view available options
1 Each
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
CAS | 1314-13-2 |
Quantity | 1 ea. |
Molecular Formula | OZn |
MDL Number | MFCD00011109 |
Synonym | zinc oxide, zinc white, amalox, permanent white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 |
InChI Key | XLOMVQKBTHCTTD-UHFFFAOYSA-N |
SMILES | O=[Zn] |
IUPAC Name | oxozinc |
Molecular Weight (g/mol) | 81.379 |
PubChem CID | 14806 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'