missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Nitrate, 6-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
154.00 € - 517.00 €
Chemical Identifiers
CAS | 10196-18-6 |
---|---|
Molecular Formula | H12N2O12Zn |
Molecular Weight (g/mol) | 297.48 |
MDL Number | MFCD00149889 |
InChI Key | JGPSMWXKRPZZRG-UHFFFAOYSA-N |
Synonym | zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a |
PubChem CID | 15865313 |
IUPAC Name | zinc;dinitrate;hexahydrate |
SMILES | O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15568284
|
Avantor J.T.Baker
4344-01 |
500 g | Poly Bottle |
154.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15578284
|
Avantor J.T.Baker
4344-05 |
2.5 kg | Poly Bottle |
517.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
10196-18-6 | |
297.48 | |
JGPSMWXKRPZZRG-UHFFFAOYSA-N | |
15865313 | |
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H12N2O12Zn | |
MFCD00149889 | |
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a | |
zinc;dinitrate;hexahydrate |
Specifications
10196-18-6 | |
H12N2O12Zn | |
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a | |
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
297.48 | |
297.47 | |
1L = 2.07kg |
Crystals | |
MFCD00149889 | |
JGPSMWXKRPZZRG-UHFFFAOYSA-N | |
zinc;dinitrate;hexahydrate | |
15865313 | |
BAKER ANALYZED™ Reagent | |
Zinc Nitrate, 6-Hydrate |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title