Learn More
Zinc meso-tetraphenylporphine, Thermo Scientific Chemicals
129.65 € - 406.02 €
Chemical Identifiers
CAS | 14074-80-7 |
---|---|
Molecular Formula | C44H28N4Zn |
Molecular Weight (g/mol) | 678.12 |
MDL Number | MFCD00012155,MFCD00012155 |
InChI Key | FGEZGTKFMCNEOZ-UHFFFAOYSA-N |
Synonym | zinc meso-tetraphenylporphine |
PubChem CID | 91667917 |
IUPAC Name | 5,10,15,20-tetraphenylporphyrin-22,24-diide;zinc |
SMILES | [Zn].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11368358
|
Thermo Scientific Alfa Aesar
039552.02 |
0.5 g |
129.65 €
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358358
|
Thermo Scientific Alfa Aesar
039552.04 |
2 g |
406.02 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A porphyrin used in light emitting diodes. It has valuable biological applications in molecular biology, fluoroimmunoassay and new pharmaceutical developments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14074-80-7 | |
678.12 | |
FGEZGTKFMCNEOZ-UHFFFAOYSA-N | |
91667917 | |
[Zn].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
C44H28N4Zn | |
MFCD00012155,MFCD00012155 | |
zinc meso-tetraphenylporphine | |
5,10,15,20-tetraphenylporphyrin-22,24-diide;zinc |
Specifications
14074-80-7 | |
Odorless | |
MFCD00012155,MFCD00012155 | |
Insoluble in water. | |
[Zn].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 | |
678.12 | |
678.1 | |
Zinc meso-tetraphenylporphine |
Purple | |
C44H28N4Zn | |
zinc meso-tetraphenylporphine | |
FGEZGTKFMCNEOZ-UHFFFAOYSA-N | |
5,10,15,20-tetraphenylporphyrin-22,24-diide;zinc | |
91667917 | |
Crystalline |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.