missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, Endotoxin Tested, Granular, Multi-Compendial, U.S.P., J.T. Baker™
359.00 € - 1005.00 €
Chemical Identifiers
CAS | 7646-85-7 |
---|---|
Molecular Formula | Cl2Zn |
Molecular Weight (g/mol) | 136.28 |
InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
Synonym | zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride |
PubChem CID | 5727 |
ChEBI | CHEBI:49976 |
IUPAC Name | dichlorozinc |
SMILES | Cl[Zn]Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15548284
|
Avantor J.T.Baker
4326-01 |
500 g | Poly Bottle |
359.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15558284
|
Avantor J.T.Baker
4326-05 |
2.5 kg | Poly Bottle |
1005.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7646-85-7 | |
136.28 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
CHEBI:49976 | |
Cl[Zn]Cl |
Cl2Zn | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
5727 | |
dichlorozinc |
Specifications
7646-85-7 | |
Cl2Zn | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
dichlorozinc | |
5727 | |
136.3 | |
1L = 2.91kg |
Granules | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
Cl[Zn]Cl | |
136.28 | |
CHEBI:49976 | |
Multi-Compendial/USP | |
Zinc Chloride, Endotoxin Tested |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zinc Chloride, Endotoxin Tested, Granular, Multi-Compendial, U.S.P., J.T. Baker™