missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, Certified AR for Analysis, Fisher Chemical™
176.00 € - 585.00 €
Chemical Identifiers
CAS | 7646-85-7 |
---|---|
Molecular Formula | Cl2Zn |
Molecular Weight (g/mol) | 136.28 |
MDL Number | 11295 |
InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
Synonym | zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride |
PubChem CID | 5727 |
ChEBI | CHEBI:49976 |
IUPAC Name | dichlorozinc |
SMILES | Cl[Zn]Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10172830
|
Fisher Chemical
Z/0870/53 |
500 g |
176.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10000540
|
Fisher Chemical
Z/0870/60 |
1 kg |
585.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7646-85-7 | |
136.28 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
5727 | |
dichlorozinc |
Cl2Zn | |
11295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
CHEBI:49976 | |
Cl[Zn]Cl |
Specifications
732°C | |
7646-85-7 | |
5 | |
Cl2Zn | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
Cl[Zn]Cl | |
136.28 | |
CHEBI:49976 | |
Plastic powder jar |
293°C | |
White | |
Solid | |
11295 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
dichlorozinc | |
5727 | |
136.29g/mol | |
1.3mbar 428 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Zinc chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title